Order Entry
Czech Republic
ContactUsLinkComponent
Osmium tetraoxide 2.5% (w/w) in tert-butanol stabilised, AcroSeal®
 
 
 
Osmium tetraoxide 2.5% (w/w) in tert-butanol stabilised, AcroSeal®
  429081000.
 :  
undefined
 
 
 
Osmium tetraoxide 2.5% (w/w) in tert-butanol stabilised, AcroSeal®
  429081000.
 :  

 

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Appearance:
    White to yellow Fused mass or solution
  • Stabilizer:
    tert-Butyl hydroperoxide
  • Strength:
    ≥2.5 wt%

 

 

 :
 :254.23 g/mol
 :130 °C (1013 hPa)
 :40 °C
 :0.811 g/cm³
 :4 °C
 :1992
 :3,II
 :GHS02
 :GHS06
 :GHS08
 :Refrigerator
 :20816-12-0
 :O₄Os
 :MFCD00011150