Order Entry
Czech Republic
ContactUsLinkComponent
Ethoxyacetylene 40% (w/w) in hexanes, AcroSeal®
Irritant
Health
Environment
Flammable
Ethoxyacetylene 40% (w/w) in hexanes, AcroSeal®
Catalog # 429061000.
CAS Number:  
undefined
Irritant
Health
Environment
Flammable
Ethoxyacetylene 40% (w/w) in hexanes, AcroSeal®
Catalog # 429061000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear orange to brown Solution
  • Identification:
    confirmed by 1H-NMR
  • NMR (1H-NMR):
    ca. 40 % w/w (typically 35 to 55 % w/w)

Specification Test Results

About this item

 :
 :70.091 g/mol
Boiling Pt:55…56 °C (760 mmHg)
Density:0.718 g/cm³
Flash Pt:–28 °C
Storage Temperature:Refrigerator
UN:1993
ADR:3,II
 :GHS07
 :GHS08
 :GHS09
 :GHS02
CAS Number:927-80-0
Formula:C₄H₆O
MDL Number:MFCD00009247