


Iodine monochloride 1 M in dichloromethane, AcroSeal®
Catalog # 428941000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Orange to red to brown Solution
- Titration with Na2S2O3:0.95 to 1.05 M
- GC:≥88 %
Specification Test Results
About this item
:
:162.36 g/mol
Density:1.42 g/cm³
Storage Temperature:Ambient
UN:2922
ADR:8,II
:GHS06
:GHS08
:GHS05
CAS Number:7790-99-0
Formula:ClI
MDL Number:MFCD00011354