
7-Methoxy-2-naphthol 97%
:5060-82-2
:
:C₁₁H₁₀O₂
:MFCD00075494
:174.2000 g/mol
:116…119 °C
:GHS07
:Ambient
- Appearance:White to brown Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:116 to 119 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Frequently Bought Together