4-Ethynylpyridine hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:352530-29-1
:
Formula:C₇H₆ClN
MDL Number:MFCD03093912
Molecular Weight:139.58 g/mol
Melting Pt:150 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to brown Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
- NMR:consistent with structure
Specification Test Results
Frequently Bought Together