
Diphenyl-2-pyridylphosphine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37943-90-1
:
Formula:C₁₇H₁₄NP
MDL Number:MFCD00192108
:263.28 g/mol
Melting Pt:82…84 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Melting point:81 to 86 °C
- Elemental Analysis:74.85 to 80.27 % C
Specification Test Results
Frequently Bought Together