Order Entry
Czech Republic
ContactUsLinkComponent
(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 97%, 98% ee
Irritant
(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 97%, 98% ee
Certificates
undefined
Irritant
(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 97%, 98% ee

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    88 to 91 °C
  • GC:
    ≥98.5 %
  • Enantiomeric excess:
    ≥97.5 %
  • Specific optical rotation:
    -118° to -122° (20 °C, 589 nm) (c=5, in CHCL3)

Specification Test Results

Related Information

Frequently Bought Together