
3,3-Diethoxy-1-propanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16777-87-0
:
Formula:C₇H₁₆O₃
MDL Number:MFCD00074850
:148.202 g/mol
Boiling Pt:90…93 °C (14 mmHg)
Density:0.941 g/cm³
Flash Pt:91 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together