Order Entry
Czech Republic
ContactUsLinkComponent
Quinine sulphate dihydrate, meets analytical specification of USP
Irritant
Quinine sulphate dihydrate, meets analytical specification of USP
Certificates
undefined
Irritant
Quinine sulphate dihydrate, meets analytical specification of USP

Specification Test Results

  • Appearance:
    White Powder or crystals or needles
  • Water:
    4.0 to 5.5 % (K.F.)
  • Heavy metals:
    ≤10 ppm
  • Identification A:
    Passes test
  • Identification B:
    Passes test
  • Identification C:
    Passes test
  • Residue after ignition:
    ≤0.1 %
  • Specific optical rotation:
    -235° to -245° (20°C, 589 nm) (c=2, 0.1 N HCl) (on anhydrous basis
  • Titration with HClO4:
    99.0 to 101.0 % (on anhydrous basis)
  • Insoluble matter:
    ≤0.1 % (in mixture of chloroform-alcohol (2:1))
  • Organic impurities:
    Passes test
  • Dihydroquinine sulfate:
    ≤10.0 %

Specification Test Results

Related Information

Frequently Bought Together