ContactUsLinkComponent
o-Cresol 99%, AcroSeal®
Toxic
Corrosive
o-Cresol 99%, AcroSeal®
Certificates
undefined
Toxic
Corrosive
o-Cresol 99%, AcroSeal®
Specification Test Results
  • Appearance:
    Colorless to amber Low melting solid or liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.2 %
  • GC:
    ≥98.5 %