
2-Bromo-5-formylpyridine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:149806-06-4
:
Formula:C₆H₄BrNO
MDL Number:MFCD04115419
:186.01 g/mol
Melting Pt:104…110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:98 to 108 °C
- GC:≥94 %
Specification Test Results
Frequently Bought Together