
D-Mannosamine hydrochloride 95%
Catalog # ACRO399170010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Specific optical rotation:-2.5° to -6.5° (20°C, 589 nm) (C=1, water)
- Titration with HClO4:≥94 %
Specification Test Results
About this item
:
:215.632 g/mol
Melting Pt:170 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:5505-63-5
:MFCD09880213
Formula:C₆H₁₄ClNO₅
MDL Number:MFCD00064557