
Methyl-4-iodo-2-methoxybenzoate 98%
:35387-93-0
:MFCD00052925
:C₉H₉IO₃
:292.07 g/mol
:93…95 °C
:GHS07
:Ambient
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:91 to 95 °C
- GC:≥97.5 %
Frequently Bought Together