Order Entry
Czech Republic
ContactUsLinkComponent
(S)-4-Hydroxypyrrolidine-2-one 98%
Irritant
(S)-4-Hydroxypyrrolidine-2-one 98%
Catalog # ACRO398970050
CAS Number:  
undefined
Irritant
(S)-4-Hydroxypyrrolidine-2-one 98%
Catalog # ACRO398970050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    156 to 159 °C
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    -40° to -46° (20 °C, 589 nm) (c=1, EtOH)

Specification Test Results

About this item

 :
 :101.105 g/mol
Melting Pt:156 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:68108-18-9
Formula:C₄H₇NO₂
MDL Number:MFCD00273370