ContactUsLinkComponent
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%
undefined
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Specific optical rotation:
    -62° to -65° (20°C, 589 nm) (C=1, acetic acid)
  • Titration with NaOH:
    ≥97.5 %
Frequently Bought Together