
3-Bromo-4-fluorobenzoic acid 98%
Catalog # ACRO397700050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:137 to 144 °C
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:219.01 g/mol
Melting Pt:138…140 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1007-16-5
Formula:C₇H₄BrFO₂
MDL Number:MFCD00042463