Order Entry
Czech Republic
ContactUsLinkComponent
 
 
4-Picolyl bromide hydrobromide 97%
 
undefined
 
4-Picolyl bromide hydrobromide 97%

 

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    189 to 192 °C
  • Titration Argentometric:
    ≥96.0 %

 

 

Frequently Bought Together