



2-Methylbutane, extra dry 99+%, AcroSeal®
:78-78-4
:
:CH₃CH₂CH(CH₃)₂
:MFCD00009338
:72.15 g/mol
:30 °C (760 mmHg)
:–160 °C
:0.62 g/cm³
:–51 °C
:1265
:3,I
:GHS07
:GHS08
:GHS09
:GHS02
:Refrigerator
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Residue after evaporation:≤0.001 %
Frequently Bought Together