Potassium acetate 99+% for molecular biology
:127-08-2
:
:CH₃COOK
:MFCD00012458
:98.14 g/mol
:292 °C
:1,57 g/cm³ (25 °C)
:>250 °C
:Ambient
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤2 %
- Titration with HClO4:≥99.0 % (On dry substance)
- Dnase,Rnase,Protease activity:None detected
- Heavy metals (as Pb):≤4 ppm
Frequently Bought Together