

1,2-Dichlorobenzene, extra dry 98+%, AcroSeal®
:95-50-1
:
:C₆H₄Cl₂
:MFCD00000535
:147 g/mol
:179…180 °C (760 mmHg)
:–15 °C
:1.3 g/cm³
:67 °C
:Ambient
:1591
:6.1,III
:GHS07
:GHS09
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥98.0 %
- Residue after evaporation:≤0.001 %
Frequently Bought Together