
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 98%
Catalog # ACRO396790250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Specific optical rotation:-2.5° to -4.5° (20°C, 589 nm) (C=4, methanol)
Specification Test Results
About this item
:
:155.6 g/mol
Melting Pt:152…161 °C
:GHS05
Storage Temperature:Refrigerator
CAS Number:5874-57-7
Formula:C₄H₉O₃N·HCl
MDL Number:MFCD00066121