ContactUsLinkComponent
1-Dodecansulphonic acid sodium salt, anhydrous 99+% for ion pair chromatography
1-Dodecansulphonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Certificates
undefined
1-Dodecansulphonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 %
  • UV:
    (0.18 M in hot water)
  • Titration after Ion exchange:
    ≥99.0 % (on dry substance)
  • Membrane filtration test:
    (on 0.18 M soln.) Passes test
  • Suitability:
    (HPLC gradient) Passes test