
Tetrabutylphosphonium hydroxide 40% (w/w) in water
Katalogové číslo ACRO395031000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Titration with HCl:37.5 to 42.5 %
Specification Test Results
About this item
:
:276.45 g/mol
Melting Pt:15 °C
Density:0.999 g/cm³
Storage Temperature:Ambient
UN:3267
ADR:8,II
:GHS05
CAS Number:14518-69-5
Formula:C₁₆H₃₇OP
MDL Number:MFCD00068456