Bismuth(III) hydroxide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10361-43-0
:
Formula:H₃BiO₃
MDL Number:MFCD00046188
Melting Pt:415 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to cream Powder
- Loss on drying:≤1.0 %
- Identification:Positive
- Bismuth (Bi):≥80.0 % (on dry basis)
Specification Test Results
Frequently Bought Together