Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
3-Furaldehyde 98% stabilised
 
undefined
 
 
3-Furaldehyde 98% stabilised

 

  • Appearance:
    Clear yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4930 to 1.4950 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Stabilizer:
    0.05 % BHT

 

 

Frequently Bought Together