
2-Bromobenzaldehyde ethylene acetal 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34824-58-3
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00155124
:229.07 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5670 to 1.5700 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together