2-(4-Bromophenyl)-1,3-dioxolane 98+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4-Bromobenzaldehyde Ethylene Acetal
- CAS Number:10602-01-4
- Formula:C₉H₉BrO₂
- MDL Number:MFCD01318950
- Molecular Weight:229.07 g/mol
- Storage Temperature:Refrigerator
2 Options Available Below
Product Details & Documents
Synonyms: 4-Bromobenzaldehyde Ethylene Acetal
- CAS Number:10602-01-4
- Formula:C₉H₉BrO₂
- MDL Number:MFCD01318950
- Molecular Weight:229.07 g/mol
- Storage Temperature:Refrigerator
Documents
ACRO39322
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Colorless Liquid or low melting solid
- Infrared spectrum:Conforms
- Refractive index:1.5630 to 1.5650 (20°C, 589 nm)
- GC:≥98.0 %



