
Triammonium citrate 97+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3458-72-8
:
Formula:C₆H₁₇N₃O₇
MDL Number:MFCD00036406
:243.22 g/mol
Melting Pt:185 °C
Density:1 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystals
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.0 %
Specification Test Results
Frequently Bought Together