
4-(N,N-Diphenylamino)benzaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4181-05-9
:
Formula:C₁₉H₁₅NO
MDL Number:MFCD00145131
Melting Pt:129…133 °C
:273.33 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:129 to 134 °C
- Water:≤2 % (K.F.)
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together