
3-(3-Pyridyl)propionic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3724-19-4
:
Formula:C₈H₉NO₂
MDL Number:MFCD00274197
:151.17 g/mol
Melting Pt:153…156 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light beige Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:153 to 157 °C
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together