
Bis(4-bromophenyl)acetylene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2789-89-1
:
Formula:C₁₄H₈Br₂
MDL Number:MFCD00185147
:336.03 g/mol
Melting Pt:183…187 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:183 to 187 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together