ContactUsLinkComponent
Silica gel 2-5 mm, non toxic for drying, with moisture indicator (orange gel)
Silica gel 2-5 mm, non toxic for drying, with moisture indicator (orange gel)
Certificates
undefined
Silica gel 2-5 mm, non toxic for drying, with moisture indicator (orange gel)
Certificates
Specification Test Results
  • Appearance:
    Orange Beads
  • Bead size:
    2 to 5 mm
  • Color:
    Orange (dry) to Colorless (wet)
  • Residual water:
    ≤2 % (150°C)
  • Water adsorption:
    ≥20 % (50% rH, 1 bar, 25°C)