
(R)-(+)-2-Methyl-2-propanesulphinamide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:196929-78-9
:
Formula:C₄H₁₁NOS
MDL Number:MFCD05861479
:121.20 g/mol
Melting Pt:99…102 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Specific optical rotation:3.0° to 5.0° (20°C, 589 nm) (c=1, chloroform)
Specification Test Results
Frequently Bought Together