1,1-Diphenyl-2-propyn-1-ol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3923-52-2
:
Formula:C₁₅H₁₂O
MDL Number:MFCD00041570
:208.26 g/mol
Boiling Pt:183 °C (20 mmHg)
Melting Pt:46…50 °C
Flash Pt:113 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to cream to light yellow Powder or crystals and/or chunks or fused solid
- Infrared spectrum:Conforms
- Melting point:44 to 50 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together