Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid ∼32% in water for analysis
Corrosive
Irritant
Hydrochloric acid ∼32% in water for analysis
Catalog # ACRO389300025
CAS Number:  
undefined
Corrosive
Irritant
Hydrochloric acid ∼32% in water for analysis
Catalog # ACRO389300025
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Residue after ign. (sulfated):
    ≤0.0005 %
  • Titration with NaOH:
    ≥32 %
  • Bromide (Br):
    ≤0.005 %
  • Sulfite (SO3):
    ≤0.0001 %
  • Sulfate (SO4):
    ≤0.0001 %
  • Aluminium (Al):
    ≤1 ppm
  • Ammonium (NH4):
    ≤0.0003 %
  • Arsenic (As):
    ≤0.1 ppm
  • Barium (Ba):
    ≤0.5 ppm
  • Cadmium (Cd):
    ≤0.01 ppm
  • Calcium (Ca):
    ≤0.5 ppm
  • Chromium (Cr):
    ≤0.02 ppm
  • Cobalt (Co):
    ≤0.01 ppm
  • Copper (Cu):
    ≤0.02 ppm
  • Iron (Fe):
    ≤0.2 ppm
  • Lead (Pb):
    ≤0.02 ppm
  • Lithium (Li):
    ≤0.02 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Manganese (Mn):
    ≤0.01 ppm
  • Nickel (Ni):
    ≤0.02 ppm
  • Potassium (K):
    ≤0.1 ppm
  • Sodium (Na):
    ≤0.5 ppm
  • Strontium (Sr):
    ≤0.02 ppm
  • Zinc (Zn):
    ≤0.05 ppm
  • Heavy metals (as Pb):
    ≤1 ppm

Specification Test Results

About this item

 :
Boiling Pt:57 °C (760 mmHg)
Melting Pt:–35 °C
Density:1.16 g/cm³
Storage Temperature:Ambient
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324