Ruthenium(VIII) oxide 0.5% in water stabilised
ACRO389000100
:
- Pack type:Glass ampoule
- Pk:10 mL
- Seq:2275
- Appearance:Clear orange to golden Solution
- Stabilizer:sodium hypochlorite
- Strength:0.5 % (typically)
:
:165.07 g/mol
:1 g/cm³
:Refrigerator
:5.1,II
:20427-56-9
:O₄Ru
:MFCD00074857