
3-Mercapto-1-propanol 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19721-22-3
:
Formula:C₃H₈OS
MDL Number:MFCD00192262
:92.16 g/mol
Boiling Pt:106 °C (50 mmHg)
Density:1.062 g/cm³
Flash Pt:85 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.4970 (20°C, 589 nm)
- GC:≥97.0 %
Specification Test Results
Frequently Bought Together