




Sodium mercury amalgam Na ∼20%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:11110-52-4
:
Formula:Hg:Na
MDL Number:MFCD00210362
Storage Temperature:Ambient
UN:3401
ADR:4.3,I
:GHS06
:GHS08
:GHS09
:GHS02
:GHS05
Specification Test Results
- Appearance:Silver to grey Irregular pieces
- Sodium (Na):20 % (approximately)
Specification Test Results
Frequently Bought Together