Order Entry
Czech Republic
ContactUsLinkComponent
1-Bromo-2-butyne 98%
Irritant
Flammable
1-Bromo-2-butyne 98%
Certificates
undefined
Irritant
Flammable
1-Bromo-2-butyne 98%

Specification Test Results

  • Appearance:
    Clear yellow-green Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5070 to 1.5090 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together