Order Entry
Czech Republic
ContactUsLinkComponent
4-(Aminomethyl)pyridine 98%
Corrosive
4-(Aminomethyl)pyridine 98%
Certificates
undefined
Corrosive
4-(Aminomethyl)pyridine 98%

Specification Test Results

  • Appearance:
    Clear colorless to yellow to orange Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5500 to 1.5530 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together