
Trimethylsulphoxonium chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5034-06-0
:
Formula:C₃H₉ClOS
MDL Number:MFCD00011898
:128.62 g/mol
Melting Pt:226…229 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
Specification Test Results
Frequently Bought Together