
trans-Dichlorobis(triethylphosphine)palladium(II)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:28425-04-9
:
Formula:C₁₂H₃₂Cl₂P₂Pd
MDL Number:MFCD00191831
:415.66 g/mol
Melting Pt:139…142 °C
Storage Temperature:Ambient
UN:3464
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Palladium (Pd):≥24.9 %
Specification Test Results
Frequently Bought Together