Order Entry
Czech Republic
ContactUsLinkComponent
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 95%
Irritant
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 95%
Catalog # ACRO370890010
CAS Number:  
undefined
Irritant
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 95%
Catalog # ACRO370890010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4450 to 1.4490 (20°C, 589 nm)
  • GC:
    ≥94 %
  • Specific optical rotation:
    +85° to +95° (20°C, 589 nm) (c=1, CHCl3)

Specification Test Results

About this item

 :
 :229.276 g/mol
Density:1.06 g/cm³
Flash Pt:107 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:95715-87-0
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD00674064