Order Entry
Czech Republic
ContactUsLinkComponent
Hydrogen chloride 1 M in acetic acid, AcroSeal®
Irritant
Corrosive
Flammable
Hydrogen chloride 1 M in acetic acid, AcroSeal®
Catalog # 370271000.
CAS Number:  
undefined
Irritant
Corrosive
Flammable
Hydrogen chloride 1 M in acetic acid, AcroSeal®
Catalog # 370271000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Strength:
    1 M Exact strength on the certificate of analysis

Specification Test Results

About this item

 :
 :36.46 g/mol
Density:1.092 g/cm³
Flash Pt:40 °C
Storage Temperature:Ambient
UN:2920
ADR:8,II
 :GHS07
 :GHS05
 :GHS02
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324