
Ethylenebis(triphenylphosphine)platinum(0) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12120-15-9
:
Formula:C₃₈H₃₄P₂Pt
MDL Number:MFCD00012028
:747.722 g/mol
Melting Pt:127…131 °C (dec.)
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:127 to 131 °C
- Assay:≥97.5 % (ex Pt)
Specification Test Results
Frequently Bought Together