

Ethyl acetate 99.8%, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141-78-6
:
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171
:88.11 g/mol
Boiling Pt:75…78 °C (760 mmHg)
Melting Pt:–83.5 °C
Density:0.902 g/cm³
Flash Pt:–4 °C
Storage Temperature:Ambient
UN:1173
ADR:3,II
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.10 % (Coulometric)
- GC:≥99.75 %
- UV:at 280 nm A: ≤0.03
- Color scale:≤10 APHA
- Specific gravity:(20°C) 0.900 to 0.904
- Residue after evaporation:≤0.0002 %
- Acidity:≤0.0008 meq/g
Specification Test Results
Related Information
Frequently Bought Together