Order Entry
Czech Republic
ContactUsLinkComponent
2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane 92%
Irritant
Flammable
2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane 92%
Certificates
undefined
Irritant
Flammable
2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane 92%

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4200 to 1.4240 (20°C, 589 nm)
  • GC:
    ≥90 %
  • Color scale:
    ≤50 APHA
  • Impurity:
    ≤0.3 % (Hydroperoxides) as 2,5-dihydroperoxy-2,5-dimethyl-hexane

Specification Test Results

Related Information

Frequently Bought Together