Zadání objednávky
Czech Republic
ContactUsLinkComponent
(S)-cis-Verbenol 97% (sum of isomers)
(S)-cis-Verbenol 97% (sum of isomers)
Certificates
undefined
(S)-cis-Verbenol 97% (sum of isomers)

Specification Test Results

  • Appearance:
    White to light yellow Crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    62 to 72 °C
  • GC:
    ≥96.0 % (sum of isomers)
  • Specific optical rotation:
    +9° to +11° (20 °C, 589 nm) (C=2, ethyl alcohol)

Specification Test Results

Related Information

Frequently Bought Together