Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Tetrafluorotetracyanoquinodimethane 95%
  ACRO312670050
 :  
undefined
 
Tetrafluorotetracyanoquinodimethane 95%
  ACRO312670050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mg
  • Seq:
    2210

 

 

  • Appearance:
    Yellow-green to orange-brown Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥94 %

 

 

 :
 :276.15 g/mol
 :291 °C (dec.)
 :Ambient
 :3439
 :6.1,III
 :GHS07
 :29261-33-4
 :C₁₂F₄N₄
 :MFCD00042382