ContactUsLinkComponent
n-Hexyllithium 33% (w/w) 2.5 N in hexane, AcroSeal®
Irritant
Health
Corrosive
Environment
Flammable
n-Hexyllithium 33% (w/w) 2.5 N in hexane, AcroSeal®
Certificates
undefined
Irritant
Health
Corrosive
Environment
Flammable
n-Hexyllithium 33% (w/w) 2.5 N in hexane, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless to yellow to light orange Solution (May develop some turbidity or precipitate)
  • Titration Active base:
    32.0 to 34.0 %
  • Free base:
    ≤1 %